Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Carboxycinnamic acid, predominantly trans, 98%

$138.54 - $138.54

Chemical Identifiers

Chemical Name or Material 4-Carboxycinnamic acid
CAS 19675-63-9
Name Note predominantly trans
Molecular Formula C10H8O4
Formula Weight 192.17
MDL Number MFCD00016543
Beilstein 2614866
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1479606

 
alfa aesar™
A1479606
5g Each for $138.54
Description & Specifications

Specifications

Formula Weight 192.17
Chemical Name or Material 4-Carboxycinnamic acid
Name Note predominantly trans
Assay 98%
Molecular Formula C10H8O4
MDL Number MFCD00016543
CAS 19675-63-9
Beilstein 2614866
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point >300°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.