missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Carboxycinnamic acid, predominantly trans, 98%

Manufacturer:  Alfa Aesar™ A1479603

 View more versions of this product

Catalog No. AAA1479603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Carboxycinnamic acid
1g
19675-63-9
MFCD00016543
4-carboxylatocinnamate, 4-e-2-carboxylatoethenyl benzoate, 4-e-3-oxido-3-oxoprop-1-enyl benzoate, 4-1e-2-carboxylatoeth-1-en-1-yl benzoate, 4-e-3-oxidanidyl-3-oxidanylidene-prop-1-enyl benzoate
C1=CC(=CC=C1C=CC(=O)[O-])C(=O)[O-]
190.154
192.17
>300°C
predominantly trans
C10H6O4-2
2614866
HAEJSGLKJYIYTB-ZZXKWVIFSA-L
4-[(E)-2-carboxylatoethenyl]benzoate
6922954
98%

Chemical Identifiers

19675-63-9
190.154
HAEJSGLKJYIYTB-ZZXKWVIFSA-L
6922954
C1=CC(=CC=C1C=CC(=O)[O-])C(=O)[O-]
C10H6O4-2
MFCD00016543
4-carboxylatocinnamate, 4-e-2-carboxylatoethenyl benzoate, 4-e-3-oxido-3-oxoprop-1-enyl benzoate, 4-1e-2-carboxylatoeth-1-en-1-yl benzoate, 4-e-3-oxidanidyl-3-oxidanylidene-prop-1-enyl benzoate
4-[(E)-2-carboxylatoethenyl]benzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

SDS Documents