missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Carboxybenzaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1527730

 View more versions of this product

Catalog No. AAA1527730



Description

Description

4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

Solubility
Soluble in water, methanol, DMSO, ether, and chloroform.

Notes
Store away from air and strong bases. Air Sensitive. Incompatible with bases and air.
Specifications

Specifications

4-Carboxybenzaldehyde
∼245°C
C8H6O3
250 g
Air Sensitive
Soluble in water,methanol,DMSO,ether,and chloroform.
C1=CC(=CC=C1C=O)C(=O)O
150.133
150.13
619-66-9
Odorless
MFCD00006951
471734
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
GOUHYARYYWKXHS-UHFFFAOYSA-N
4-formylbenzoic acid
12088
98%

Chemical Identifiers

619-66-9
150.133
GOUHYARYYWKXHS-UHFFFAOYSA-N
12088
C1=CC(=CC=C1C=O)C(=O)O
C8H6O3
MFCD00006951
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
4-formylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c

H315-H319-H335-H373

EINECSNumber : 210-607-4

RTECSNumber : WZ0440000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only