missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Carboxybenzaldehyde, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC154585000

 View more versions of this product

Catalog No. AC154585000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Carboxybenzaldehyde
Glass bottle
247°C
619-66-9
100.0
C8H6O3
MFCD00006951
4-carboxybenzaladehyde
GOUHYARYYWKXHS-UHFFFAOYSA-N
4-formylbenzoic acid
12088
Fine Crystalline Powder
Authentic
Yellow
500g
95.0
95% min. (HPLC)
HO2CC6H4CHO
10, 671
Solubility in water: soluble
C1=CC(=CC=C1C=O)C(=O)O
150.13
150.13
96%

Chemical Identifiers

619-66-9
150.13
GOUHYARYYWKXHS-UHFFFAOYSA-N
12088
C1=CC(=CC=C1C=O)C(=O)O
C8H6O3
MFCD00006951
4-carboxybenzaladehyde
4-formylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-607-4

SDS Documents