Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Carboxy-3-chlorobenzeneboronic acid, 97%

$86.66 - $250.91

Chemical Identifiers

Chemical Name or Material 4-Carboxy-3-chlorobenzeneboronic acid
CAS 136496-72-5
Molecular Formula C7H6BClO4
Formula Weight 200.39
MDL Number MFCD06656268
Synonym 4-Borono-2-chlorobenzoic acid; 4-Carboxy-3-chlorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2899906

 
alfa aesar™
H2899906
5g Each for $250.91

AAH2899903

 
alfa aesar™
H2899903
1g Each for $86.66
Description & Specifications

Specifications

Formula Weight 200.39
Chemical Name or Material 4-Carboxy-3-chlorobenzeneboronic acid
Assay 97%
Molecular Formula C7H6BClO4
MDL Number MFCD06656268
CAS 136496-72-5
Synonym 4-Borono-2-chlorobenzoic acid; 4-Carboxy-3-chlorophenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.