missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6202806

 View more versions of this product

Catalog No. AAH6202806



Specifications

Specifications

4-Carboxy-2-fluorobenzeneboronic acid pinacol ester
5g
C13H16BFO4
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)O)F
266.075
266.07
96%
199°C to 204°C
1050423-87-4
MFCD12756466
JSQNZNXVVRPGTD-UHFFFAOYSA-N
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
46739808
Solid

Chemical Identifiers

1050423-87-4
266.075
JSQNZNXVVRPGTD-UHFFFAOYSA-N
46739808
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)O)F
C13H16BFO4
MFCD12756466
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents