Alfa Aesar™ 4-Carboxy-2-fluorobenzeneboronic acid, 97%
Manufacturer:  Alfa Aesar™ H55857MD
 View more versions of this product
Catalog No. AAH55857MD
Specifications
Specifications
4-Carboxy-2-fluorobenzeneboronic acid
851135-07-4
MFCD08458473
TVNVDRMGOOLVOX-UHFFFAOYSA-N
4-borono-3-fluorobenzoic acid
23134177
97%
250mg
C7H6BFO4
3-fluoro-4-dihydroxyboryl benzoic acid
B(C1=C(C=C(C=C1)C(=O)O)F)(O)O
183.929
183.93
Chemical Identifiers
851135-07-4
183.929
TVNVDRMGOOLVOX-UHFFFAOYSA-N
23134177
B(C1=C(C=C(C=C1)C(=O)O)F)(O)O
C7H6BFO4
MFCD08458473
3-fluoro-4-dihydroxyboryl benzoic acid
4-borono-3-fluorobenzoic acid
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
Documents