missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Carbamoyltetrahydropyran, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450880010
Specifications
4-Carbamoyltetrahydropyran | |
180.0°C to 182.0°C | |
344329-76-6 | |
C6H11NO2 | |
DGOYLVBDCVINQZ-UHFFFAOYSA-N | |
oxane-4-carboxamide | |
13197203 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
2h-3,4,5,6-tetrahydropyran-4-carboxamide, 2h-pyran-4-carboxamide, tetrahydro, 2h-pyran-4-carboxamide,tetrahydro, 4-carbamoyltetrahydro-2h-pyran, 4-carbamoyltetrahydropyran, acmc-209i7n, pubchem9735, tetrahydro-2h-4-pyrancarboxamide, tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide | |
C1COCCC1C(=O)N | |
129.16 | |
129.16 |
Chemical Identifiers
344329-76-6 | |
129.16 | |
2h-3,4,5,6-tetrahydropyran-4-carboxamide, 2h-pyran-4-carboxamide, tetrahydro, 2h-pyran-4-carboxamide,tetrahydro, 4-carbamoyltetrahydro-2h-pyran, 4-carbamoyltetrahydropyran, acmc-209i7n, pubchem9735, tetrahydro-2h-4-pyrancarboxamide, tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide | |
oxane-4-carboxamide |
C6H11NO2 | |
DGOYLVBDCVINQZ-UHFFFAOYSA-N | |
13197203 | |
C1COCCC1C(=O)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning