missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Carbamoyltetrahydropyran, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450880050

 View more versions of this product

Catalog No. AC450880050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Carbamoyltetrahydropyran
5g
96% min. (GC)
2h-3,4,5,6-tetrahydropyran-4-carboxamide, 2h-pyran-4-carboxamide, tetrahydro, 2h-pyran-4-carboxamide,tetrahydro, 4-carbamoyltetrahydro-2h-pyran, 4-carbamoyltetrahydropyran, acmc-209i7n, pubchem9735, tetrahydro-2h-4-pyrancarboxamide, tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide
C1COCCC1C(=O)N
129.16
129.16
180.0°C to 182.0°C
344329-76-6
C6H11NO2
DGOYLVBDCVINQZ-UHFFFAOYSA-N
oxane-4-carboxamide
13197203
97%

Chemical Identifiers

344329-76-6
129.16
2h-3,4,5,6-tetrahydropyran-4-carboxamide, 2h-pyran-4-carboxamide, tetrahydro, 2h-pyran-4-carboxamide,tetrahydro, 4-carbamoyltetrahydro-2h-pyran, 4-carbamoyltetrahydropyran, acmc-209i7n, pubchem9735, tetrahydro-2h-4-pyrancarboxamide, tetrahydro-2h-pyran-4-carboxamide, tetrahydropyran-4-carboxamide
oxane-4-carboxamide
C6H11NO2
DGOYLVBDCVINQZ-UHFFFAOYSA-N
13197203
C1COCCC1C(=O)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents