missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Carbamoyl-3-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5487703

 View more versions of this product

Catalog No. AAH5487703



Specifications

Specifications

4-Carbamoyl-3-fluorobenzeneboronic acid
874288-39-8
MFCD09258667
LKXXEWDYAWOZRW-UHFFFAOYSA-N
(4-carbamoyl-3-fluorophenyl)boronic acid
44717670
97%
1g
C7H7BFNO3
4-aminocarbonyl-3-fluoranyl-phenyl boronic acid
B(C1=CC(=C(C=C1)C(=O)N)F)(O)O
182.945
182.95

Chemical Identifiers

874288-39-8
182.945
LKXXEWDYAWOZRW-UHFFFAOYSA-N
44717670
B(C1=CC(=C(C=C1)C(=O)N)F)(O)O
C7H7BFNO3
MFCD09258667
4-aminocarbonyl-3-fluoranyl-phenyl boronic acid
(4-carbamoyl-3-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents