missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Butylcyclohexanol (cis- and trans- mixture), TCI America™

$198.61 - $640.16

Chemical Identifiers

CAS 70568-60-4
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD01321147
InChI Key NUANGSLQWFBVEH-UHFFFAOYSA-N
PubChem CID 4132186
IUPAC Name 4-butylcyclohexan-1-ol
SMILES CCCCC1CCC(CC1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19305G View Documents TCI America
B19305G
5g Each for $198.61
 
B193025G View Documents TCI America
B193025G
25g Each for $640.16
 
Specifications

Chemical Identifiers

70568-60-4
156.269
NUANGSLQWFBVEH-UHFFFAOYSA-N
4-butylcyclohexan-1-ol
C10H20O
MFCD01321147
4132186
CCCCC1CCC(CC1)O

Specifications

122°C
5g
C10H20O
NUANGSLQWFBVEH-UHFFFAOYSA-N
4-butylcyclohexan-1-ol
4132186
Clear Liquid at 20°C
4-Butylcyclohexanol (cis- and trans- mixture)
Colorless
70568-60-4
MFCD01321147
CCCCC1CCC(CC1)O
156.269
156.27
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2301

TSCA : No