Please login to your online account to display your discounted pricing

4-Butylbenzene-1-sulfonyl chloride, 97%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  BTB10243EB

 View more versions of this product

Catalog No. BTB10243EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.73
Quantity 25g
Assay 97%
CAS Max % 100.0
Linear Formula CH3(CH2)3C6H4SO2Cl
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cau
CAS 54997-92-1
Chemical Name or Material 4-Butylbenzene-1-sulfonyl chloride
MDL Number MFCD00173908
CAS Min % 96.5
Health Hazard 1 Danger
Assay Percent Range 97%
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
Molecular Formula C10H13ClO2S

Specifications

Chemical Name or Material 4-Butylbenzene-1-sulfonyl chloride
CAS 54997-92-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13ClO2S
Linear Formula CH3(CH2)3C6H4SO2Cl
Formula Weight 232.73
MDL Number MFCD00173908
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cau

GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.

SDS & Safety Documents