missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Butylbenzaldehyde Diethyl Acetal, TCI America™

$120.70 - $363.63

Chemical Identifiers

CAS 83803-80-9
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00009466
InChI Key VZIJDIWCWDYVFM-UHFFFAOYSA-N
Synonym 1-butyl-4-diethoxymethyl benzene, 4-butylbenzaldehyde diethylacetal, 4-n-butylbenzaldehyde diethyl acetal, 4-butylbenzaldehyde diethyl acetal, 1-butyl-4-diethoxymethyl-benzene, benzene,1-butyl-4-diethoxymethyl, 4-butylphenyl diethoxymethane, 4-butylbenzaldehydediethylacetal, acmc-209ptb, 4-butyl-, a,, a-diethoxytoluene
PubChem CID 605679
IUPAC Name 1-butyl-4-(diethoxymethyl)benzene
SMILES CCCCC1=CC=C(C=C1)C(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12225ML View Documents TCI America
B12225ML
5mL Each for $120.70
 
B122225ML View Documents TCI America
B122225ML
25mL Each for $363.63
 
Specifications

Chemical Identifiers

83803-80-9
236.355
VZIJDIWCWDYVFM-UHFFFAOYSA-N
605679
CCCCC1=CC=C(C=C1)C(OCC)OCC
C15H24O2
MFCD00009466
1-butyl-4-diethoxymethyl benzene, 4-butylbenzaldehyde diethylacetal, 4-n-butylbenzaldehyde diethyl acetal, 4-butylbenzaldehyde diethyl acetal, 1-butyl-4-diethoxymethyl-benzene, benzene,1-butyl-4-diethoxymethyl, 4-butylphenyl diethoxymethane, 4-butylbenzaldehydediethylacetal, acmc-209ptb, 4-butyl-, a,, a-diethoxytoluene
1-butyl-4-(diethoxymethyl)benzene

Specifications

Yellow
83803-80-9
MFCD00009466
VZIJDIWCWDYVFM-UHFFFAOYSA-N
1-butyl-4-(diethoxymethyl)benzene
605679
Clear Liquid at 20°C
4-Butylbenzaldehyde Diethyl Acetal
5mL
C15H24O2
1-butyl-4-diethoxymethyl benzene, 4-butylbenzaldehyde diethylacetal, 4-n-butylbenzaldehyde diethyl acetal, 4-butylbenzaldehyde diethyl acetal, 1-butyl-4-diethoxymethyl-benzene, benzene,1-butyl-4-diethoxymethyl, 4-butylphenyl diethoxymethane, 4-butylbenzaldehydediethylacetal, acmc-209ptb, 4-butyl-, a,, a-diethoxytoluene
CCCCC1=CC=C(C=C1)C(OCC)OCC
236.355
236.36
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No