missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Butoxybenzyl Alcohol, TCI America™

$84.03 - $284.17

Chemical Identifiers

CAS 6214-45-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD00004657
InChI Key MGKNBDBZIKPUFM-UHFFFAOYSA-N
Synonym 4-butoxybenzyl alcohol, 4-butoxyphenyl methanol, 4-n-butoxybenzyl alcohol, p-butoxybenzyl alcohol, benzenemethanol, 4-butoxy, 4-butoxybenzylalcohol, benzenemethanol,4-butoxy, 4-butoxy-phenyl-methanol, 4-butoxyphenyl methanol #, acmc-1b65d
PubChem CID 80346
IUPAC Name (4-butoxyphenyl)methanol
SMILES CCCCOC1=CC=C(C=C1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B10425G View Documents TCI America
B10425G
5g Each for $84.03
 
B104225G View Documents TCI America
B104225G
25g Each for $284.17
 
Specifications

Chemical Identifiers

6214-45-5
180.247
MGKNBDBZIKPUFM-UHFFFAOYSA-N
80346
CCCCOC1=CC=C(C=C1)CO
C11H16O2
MFCD00004657
4-butoxybenzyl alcohol, 4-butoxyphenyl methanol, 4-n-butoxybenzyl alcohol, p-butoxybenzyl alcohol, benzenemethanol, 4-butoxy, 4-butoxybenzylalcohol, benzenemethanol,4-butoxy, 4-butoxy-phenyl-methanol, 4-butoxyphenyl methanol #, acmc-1b65d
(4-butoxyphenyl)methanol

Specifications

White-Yellow
6214-45-5
MFCD00004657
MGKNBDBZIKPUFM-UHFFFAOYSA-N
(4-butoxyphenyl)methanol
80346
Crystal-Lump at 20°C
4-Butoxybenzyl Alcohol
5g
C11H16O2
4-butoxybenzyl alcohol, 4-butoxyphenyl methanol, 4-n-butoxybenzyl alcohol, p-butoxybenzyl alcohol, benzenemethanol, 4-butoxy, 4-butoxybenzylalcohol, benzenemethanol,4-butoxy, 4-butoxy-phenyl-methanol, 4-butoxyphenyl methanol #, acmc-1b65d
CCCCOC1=CC=C(C=C1)CO
180.247
180.25
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No