Please login to your online account to display your discounted pricing

4-Butoxybenzene-1-sulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Butoxybenzene-1-sulfonyl chloride
CAS 1138-56-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13ClO3S
Linear Formula CH3(CH2)3OC6H4SO2Cl
Formula Weight 248.73
MDL Number MFCD00052344
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00441EA

  SDS
fisher scientific company
TL00441EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.73
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Assay Percent Range 97%
MDL Number MFCD00052344
Linear Formula CH3(CH2)3OC6H4SO2Cl
CAS 1138-56-3
Chemical Name or Material 4-Butoxybenzene-1-sulfonyl chloride
Molecular Formula C10H13ClO3S