Please login to your online account to display your discounted pricing

4-Butoxybenzaldehyde, 99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218370500

 View more versions of this product

Catalog No. AC218370500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Color Clear yellow to orang-red
Formula Weight 178.23
Physical Form Liquid
Quantity 50g
Beilstein 08, II, 67
CAS Max % 100.0
Linear Formula CH3(CH2)3OC6H4CHO
CAS 5736-88-9
Molecular Formula C11H14O2
Assay Percent Range 99%
MDL Number MFCD00003389
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 4-Butoxybenzaldehyde

Specifications

Chemical Name or Material 4-Butoxybenzaldehyde
CAS 5736-88-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H14O2
Linear Formula CH3(CH2)3OC6H4CHO
Formula Weight 178.23
MDL Number MFCD00003389