missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Butoxyaniline, TCI America™

$36.67 - $119.17

Chemical Identifiers

CAS 4344-55-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00007866
InChI Key UBRIHZOFEJHMIT-UHFFFAOYSA-N
Synonym p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
PubChem CID 20352
IUPAC Name 4-butoxyaniline
SMILES CCCCOC1=CC=C(C=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B10035G View Documents TCI America
B10035G
5g Each for $36.67
 
B100325G View Documents TCI America
B100325G
25g Each for $119.17
 
Specifications

Chemical Identifiers

4344-55-2
165.236
UBRIHZOFEJHMIT-UHFFFAOYSA-N
20352
CCCCOC1=CC=C(C=C1)N
C10H15NO
MFCD00007866
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
4-butoxyaniline

Specifications

157°C
5g
C10H15NO
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
CCCCOC1=CC=C(C=C1)N
165.236
165.24
≥98.0% (GC,T)
Brown
4344-55-2
MFCD00007866
UBRIHZOFEJHMIT-UHFFFAOYSA-N
4-butoxyaniline
20352
Clear Liquid at 20°C
4-Butoxyaniline
Safety and Handling

Safety and Handling

TSCA : Yes