Please login to your online account to display your discounted pricing

4-Butoxyaniline, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  293260250

 View more versions of this product

Catalog No. AC293260250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 165.24
Quantity 25g
Color Clear dark brown
EINECS Number 224-402-2
TSCA TSCA
RTECS Number BW9420000
Beilstein 13, II, 226
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material 4-Butoxyaniline
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H15NO
Refractive Index 1.5365 to 1.5395 (20°C, 589nm)
Linear Formula CH3(CH2)3OC6H4NH2
MDL Number MFCD00007866
CAS 4344-55-2
Infrared Spectrum Authentic
Synonym 4-Butyloxybenzenamine
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Butoxyaniline
Name Note 97%
CAS 4344-55-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H15NO
Linear Formula CH3(CH2)3OC6H4NH2
Formula Weight 165.24
MDL Number MFCD00007866
Synonym 4-Butyloxybenzenamine
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents