missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Butoxyaniline, 97%, ACROS Organics™

$280.11 - $280.11

Chemical Identifiers

CAS 4344-55-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H15NO
Linear Formula CH3(CH2)3OC6H4NH2
Formula Weight 165.24
MDL Number MFCD00007866
Synonym 4-Butyloxybenzenamine
Chemical Name or Material 4-Butoxyaniline, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC293260250 SDS Thermo Fisher Scientific
293260250
25g Each for $280.11
Specifications Description & Specifications

Chemical Identifiers

4344-55-2
100.0
C10H15NO
165.24
4-Butyloxybenzenamine
≥96.0%
96.0
97%
CH3(CH2)3OC6H4NH2
MFCD00007866
4-Butoxyaniline, 97%

Specifications

Liquid
25g
13, II, 226
96.0
CH3(CH2)3OC6H4NH2
4344-55-2
97%
4-Butyloxybenzenamine
≥96.0%
165.24
Clear dark brown
100.0
1.5365 to 1.5395 (20°C, 589nm)
C10H15NO
MFCD00007866
Authentic
4-Butoxyaniline, 97%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 224-402-2

RTECSNumber : BW9420000

TSCA : TSCA