missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Butoxy-4'-cyanobiphenyl 98.0+%, TCI America™
$227.29 - $791.35
Chemical Identifiers
CAS | 52709-87-2 |
---|---|
Molecular Formula | C17H17NO |
Molecular Weight (g/mol) | 251.33 |
MDL Number | MFCD00183755 |
InChI Key | KPQVQWUNELODQE-UHFFFAOYSA-N |
PubChem CID | 104292 |
IUPAC Name | 4'-butoxy-[1,1'-biphenyl]-4-carbonitrile |
SMILES | CCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N |
Chemical Identifiers
52709-87-2 | |
251.33 | |
KPQVQWUNELODQE-UHFFFAOYSA-N | |
4'-butoxy-[1,1'-biphenyl]-4-carbonitrile |
C17H17NO | |
MFCD00183755 | |
104292 | |
CCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N |
Specifications
52709-87-2 | |
White | |
MFCD00183755 | |
4′C-Butyloxy-4-biphenylcarbonitrile, 4OCB | |
CCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N | |
251.33 | |
251.33 | |
Crystalline Powder |
77°C | |
C17H17NO | |
5 g | |
KPQVQWUNELODQE-UHFFFAOYSA-N | |
4'-butoxy-[1,1'-biphenyl]-4-carbonitrile | |
104292 | |
≥98.0% (GC) | |
4-Butoxy-4′-cyanobiphenyl |
Safety and Handling
RTECSNumber : DV1764000
TSCA : Yes