Please login to your online account to display your discounted pricing

4-Bromoveratrole 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  107540050

 View more versions of this product

Catalog No. AC107540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 217.06
Physical Form Liquid
Quantity 5g
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 220-677-8
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5720 to 1.5740 (20°C, 589nm)
Chemical Name or Material 4-Bromoveratrole
Linear Formula BrC6H3(OCH3)2
CAS 2859-78-1
MDL Number MFCD00008381
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H9BrO2
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4-Bromoveratrole
Name Note 97%
CAS 2859-78-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9BrO2
Linear Formula BrC6H3(OCH3)2
Formula Weight 217.06
MDL Number MFCD00008381
Accessories

Warning

SDS & Safety Documents