Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromovalerophenone, 98%

$39.93 - $476.14

Chemical Identifiers

Chemical Name or Material 4'-Bromovalerophenone
CAS 7295-44-5
Molecular Formula C11H13BrO
Formula Weight 241.14
MDL Number MFCD00000107
Synonym 4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2497906

  SDS
alfa aesar™
B2497906
5g Each for $39.93

AAB2497914

  SDS
alfa aesar™
B2497914
25g Each for $139.99

AAB2497922

  SDS
alfa aesar™
B2497922
100g Each for $476.14
Description & Specifications

Specifications

Formula Weight 241.14
Chemical Name or Material 4'-Bromovalerophenone
Boiling Point 168° to 169°C (20mmHg)
MDL Number MFCD00000107
Melting Point 34° to 36°C
Synonym 4-Bromphenyl n-butyl ketone; 1-(4-Bromophenyl)pentan-1-one
Molecular Formula C11H13BrO
Flash Point >110°C (230°F)
CAS 7295-44-5
Quantity 5g
Assay 98%