missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromotriphenylamine, 99%

$109.35 - $445.68

Chemical Identifiers

CAS 36809-26-4
Molecular Formula C18H14BrN
Molecular Weight (g/mol) 324.221
MDL Number MFCD01851266
InChI Key SQTLUXJWUCHKMT-UHFFFAOYSA-N
Synonym 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, 4-bromophenyl diphenylamine, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
PubChem CID 642838
IUPAC Name 4-bromo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6032806 View Documents Alfa Aesar™
H6032806
5g Each for $109.35
 
AAH6032814 View Documents Alfa Aesar™
H6032814
25g Each for $445.68
 
Specifications

Chemical Identifiers

36809-26-4
324.221
SQTLUXJWUCHKMT-UHFFFAOYSA-N
642838
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br
C18H14BrN
MFCD01851266
4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, 4-bromophenyl diphenylamine, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
4-bromo-N,N-diphenylaniline

Specifications

108°C to 112°C
36809-26-4
MFCD01851266
SQTLUXJWUCHKMT-UHFFFAOYSA-N
4-bromo-N,N-diphenylaniline
642838
99%
5g
C18H14BrN
4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, 4-bromophenyl diphenylamine, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br
324.221
324.21
4-Bromotriphenylamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger