Please login to your online account to display your discounted pricing

4-Bromothiophenol, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107460050

 View more versions of this product

Catalog No. AC107460050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Assay ≥94%
Formula Weight 189.07
Quantity 5g
Color White to beige
EINECS Number 203-407-3
TSCA TSCA
Melting Point 71° to 76°C
Beilstein 06, 330
CAS Max % 100.0
CAS Min % 94.0
Synonym 4-Bromobenzenethiol
Chemical Name or Material 4-Bromothiophenol
Name Note 95%
Assay Percent Range 95%
Molecular Formula C6H5BrS
Linear Formula BrC6H4SH
CAS 106-53-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic if swallowed.
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00004845
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Bromothiophenol
Name Note 95%
CAS 106-53-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5BrS
Linear Formula BrC6H4SH
Formula Weight 189.07
MDL Number MFCD00004845
Synonym 4-Bromobenzenethiol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic if swallowed.

SDS & Safety Documents