missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5595606

 View more versions of this product

Catalog No. AAH5595606



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromothiobenzamide
139°C to 143°C
C7H6BrNS
5 g
AIPANIYQEBQYGC-UHFFFAOYSA-N
4-bromobenzenecarbothioamide
1549289
97%
26197-93-3
Brown to White
MFCD04114360
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
C1=CC(=CC=C1C(=S)N)Br
216.096
216.1
Solid

Chemical Identifiers

26197-93-3
216.096
AIPANIYQEBQYGC-UHFFFAOYSA-N
1549289
C1=CC(=CC=C1C(=S)N)Br
C7H6BrNS
MFCD04114360
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
4-bromobenzenecarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only