Please login to your online account to display your discounted pricing

4-Bromothioanisole, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188190050

 View more versions of this product

Catalog No. AC188190050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 203.1
Assay ≥96.0%
Physical Form Low Melting Crystalline Mass, Crystals, Crystalline Powder or Chunks
Quantity 5g
Color White to beige
Melting Point 35° to 40°C
EINECS Number 203-255-8
Beilstein 06, 330
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Name Note 97%
Linear Formula BrC6H4SCH3
MDL Number MFCD00000102
Assay Percent Range 97%
Chemical Name or Material 4-Bromothioanisole
Infrared Spectrum Authentic
CAS 104-95-0
Molecular Formula C7H7BrS
Synonym 4-Bromophenyl methyl sulfide
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Bromothioanisole
Name Note 97%
CAS 104-95-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7BrS
Linear Formula BrC6H4SCH3
Formula Weight 203.1
MDL Number MFCD00000102
Synonym 4-Bromophenyl methyl sulfide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents