missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromothioanisole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 188191000

 View more versions of this product

Catalog No. AC188191000



Description

Description

Specifications

Specifications

4-Bromothioanisole
Authentic
White to Beige
100g
104-95-0
100.0
C7H7BrS
MFCD00000102
1-bromo-4-methylsulfanyl benzene, 1-bromo-4-methylthio benzene, 4-bromo-thioanisole, 4-bromophenyl methyl sulfide, 4-bromothioanisole, benzene, 1-bromo-4-methylthio, methyl 4-bromophenylsulfide, p-bromo thioanisole, p-bromophenyl methyl sulfide, p-bromothioanisole
CSC1=CC=C(C=C1)Br
203.1
203.1
97%
>110°C
Glass bottle
35.0°C to 40.0°C
97%
96.0
96% min. (GC)
BrC6H4SCH3
06, 330
YEUYZNNBXLMFCW-UHFFFAOYSA-N
1-bromo-4-methylsulfanylbenzene
66037
Low Melting Crystalline Mass, Crystals, Crystalline Powder or Chunks

Chemical Identifiers

104-95-0
203.1
YEUYZNNBXLMFCW-UHFFFAOYSA-N
66037
CSC1=CC=C(C=C1)Br
C7H7BrS
MFCD00000102
1-bromo-4-methylsulfanyl benzene, 1-bromo-4-methylthio benzene, 4-bromo-thioanisole, 4-bromophenyl methyl sulfide, 4-bromothioanisole, benzene, 1-bromo-4-methylthio, methyl 4-bromophenylsulfide, p-bromo thioanisole, p-bromophenyl methyl sulfide, p-bromothioanisole
1-bromo-4-methylsulfanylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 203-255-8

SDS Documents