Learn More
4-Bromothiazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 429450010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Bromothiazole | |
96.0 | |
102°C | |
96% min. (GC) | |
C3H2BrNS | |
1g | |
VDTIGYKLTROQAH-UHFFFAOYSA-N | |
4-bromo-1,3-thiazole | |
2763218 | |
97% |
34259-99-9 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD06657592 | |
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h | |
BrC1=CSC=N1 | |
164.02 | |
164.02 |
Chemical Identifiers
34259-99-9 | |
164.02 | |
VDTIGYKLTROQAH-UHFFFAOYSA-N | |
2763218 | |
BrC1=CSC=N1 |
C3H2BrNS | |
MFCD06657592 | |
4-bromothiazole, 4-bromo-thiazole, 4-bromo thiazole, thiazole, 4-bromo, 4-bromothiazol, zlchem 498, pubchem4040, ksc222g1h | |
4-bromo-1,3-thiazole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
RUO â Research Use Only