missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromotetraphenylsilane, TCI America™

$268.89 - $870.84

Chemical Identifiers

CAS 18737-40-1
Molecular Formula C24H19BrSi
Molecular Weight (g/mol) 415.405
InChI Key UDZSLJULKCKKPX-UHFFFAOYSA-N
PubChem CID 297506
IUPAC Name (4-bromophenyl)-triphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B3287200MG View Documents TCI America
B3287200MG
200mg Each for $268.89
 
B32871G View Documents TCI America
B32871G
1g Each for $870.84
 
Specifications

Chemical Identifiers

18737-40-1
415.405
297506
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br
C24H19BrSi
UDZSLJULKCKKPX-UHFFFAOYSA-N
(4-bromophenyl)-triphenylsilane

Specifications

White-Yellow
200mg
C24H19BrSi
C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br
415.405
415.41
≥96.0% (GC)
175°C
18737-40-1
UDZSLJULKCKKPX-UHFFFAOYSA-N
(4-bromophenyl)-triphenylsilane
297506
Crystal-Powder at 20°C
4-Bromotetraphenylsilane
Safety and Handling

Safety and Handling

TSCA : No