Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-4-Bromostyrene oxide, 98%

Chemical Identifiers

Chemical Name or Material (+/-)-4-Bromostyrene oxide
CAS 32017-76-8
Molecular Formula C8H7BrO
Formula Weight 199.04
MDL Number MFCD01656787
UN Number UN1325
Synonym 2-(4-Bromophenyl)oxirane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2681806

 
alfa aesar™
H2681806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2681814

 
alfa aesar™
H2681814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.04
Chemical Name or Material (+/-)-4-Bromostyrene oxide
Assay 98%
Molecular Formula C8H7BrO
MDL Number MFCD01656787
CAS 32017-76-8
UN Number UN1325
Melting Point 26° to 29°C
Synonym 2-(4-Bromophenyl)oxirane
Boiling Point 91° to 93°C (2mmHg)
Quantity 5g
Flash Point 38°C (100°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H341-H315-H319-H335
Flammable solid.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECS Number CY9025000

Applications
(+/-)-4-Bromostyrene oxide is used for the synthesis of pharmaceuticals, disinfectants, and products for the perfume industry.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H341-H315-H319-H335
Flammable solid.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.