missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromopyridine Hydrobromide, TCI America™

$82.50 - $247.50

Chemical Identifiers

CAS 74129-11-6
Molecular Formula C5H5Br2N
Molecular Weight (g/mol) 238.91
MDL Number MFCD00060195
InChI Key SMSVBXDZZQOTGB-UHFFFAOYSA-N
PubChem CID 44630127
IUPAC Name 4-bromopyridine;hydrobromide
SMILES C1=CN=CC=C1Br.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11715G View Documents TCI America
B11715G
5g Each for $82.50
 
B117125G View Documents TCI America
B117125G
25g Each for $247.50
 
Specifications

Chemical Identifiers

74129-11-6
238.91
SMSVBXDZZQOTGB-UHFFFAOYSA-N
4-bromopyridine;hydrobromide
C5H5Br2N
MFCD00060195
44630127
C1=CN=CC=C1Br.Br

Specifications

White-Yellow
74129-11-6
MFCD00060195
SMSVBXDZZQOTGB-UHFFFAOYSA-N
4-bromopyridine;hydrobromide
44630127
Crystal-Powder at 20°C
4-Bromopyridine Hydrobromide
5g
C5H5Br2N
2811
C1=CN=CC=C1Br.Br
238.91
238.91
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator