missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromopyridine-3-carboxylic acid, 96%

$119.05 - $454.92

Chemical Identifiers

CAS 15366-62-8
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD04114236
InChI Key CZLOEKRJQIAKFI-UHFFFAOYSA-N
Synonym 4-bromonicotinic acid, 4-bromo-nicotinic acid, 3-acid-4-bromopyridine, 3-pyridinecarboxylic acid, 4-bromo, 4-bromo-nicotinicacid, 4-bromo-3-pyridinecarboxylic acid, 4-bromonicotinicacid, 3-acid-4-bromopyridine, pubchem18140, acmc-209d9s
PubChem CID 2762915
IUPAC Name 4-bromopyridine-3-carboxylic acid
SMILES C1=CN=CC(=C1Br)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6632203 View Documents Alfa Aesar™
H6632203
1g Each for $119.05
 
AAH6632206 View Documents Alfa Aesar™
H6632206
5g Each for $454.92
 
Specifications

Chemical Identifiers

15366-62-8
202.007
CZLOEKRJQIAKFI-UHFFFAOYSA-N
2762915
C1=CN=CC(=C1Br)C(=O)O
C6H4BrNO2
MFCD04114236
4-bromonicotinic acid, 4-bromo-nicotinic acid, 3-acid-4-bromopyridine, 3-pyridinecarboxylic acid, 4-bromo, 4-bromo-nicotinicacid, 4-bromo-3-pyridinecarboxylic acid, 4-bromonicotinicacid, 3-acid-4-bromopyridine, pubchem18140, acmc-209d9s
4-bromopyridine-3-carboxylic acid

Specifications

Brown
1g
C6H4BrNO2
4-bromonicotinic acid, 4-bromo-nicotinic acid, 3-acid-4-bromopyridine, 3-pyridinecarboxylic acid, 4-bromo, 4-bromo-nicotinicacid, 4-bromo-3-pyridinecarboxylic acid, 4-bromonicotinicacid, 3-acid-4-bromopyridine, pubchem18140, acmc-209d9s
C1=CN=CC(=C1Br)C(=O)O
202.007
202
96%
118°C to 134°C
15366-62-8
MFCD04114236
CZLOEKRJQIAKFI-UHFFFAOYSA-N
4-bromopyridine-3-carboxylic acid
2762915
Solid
4-Bromopyridine-3-carboxylic acid