missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromopyridine-3-carboxylic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6632206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromopyridine-3-carboxylic acid | |
118°C to 134°C | |
C6H4BrNO2 | |
5 g | |
CZLOEKRJQIAKFI-UHFFFAOYSA-N | |
4-bromopyridine-3-carboxylic acid | |
2762915 | |
96% |
15366-62-8 | |
Brown | |
MFCD04114236 | |
4-bromonicotinic acid, 4-bromo-nicotinic acid, 3-acid-4-bromopyridine, 3-pyridinecarboxylic acid, 4-bromo, 4-bromo-nicotinicacid, 4-bromo-3-pyridinecarboxylic acid, 4-bromonicotinicacid, pubchem18140, acmc-209d9s | |
OC(=O)C1=C(Br)C=CN=C1 | |
202.01 | |
202 | |
Solid |
Chemical Identifiers
15366-62-8 | |
202.01 | |
CZLOEKRJQIAKFI-UHFFFAOYSA-N | |
2762915 | |
OC(=O)C1=C(Br)C=CN=C1 |
C6H4BrNO2 | |
MFCD04114236 | |
4-bromonicotinic acid, 4-bromo-nicotinic acid, 3-acid-4-bromopyridine, 3-pyridinecarboxylic acid, 4-bromo, 4-bromo-nicotinicacid, 4-bromo-3-pyridinecarboxylic acid, 4-bromonicotinicacid, pubchem18140, acmc-209d9s | |
4-bromopyridine-3-carboxylic acid |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only