missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromopyridine-2-carboxylic acid, 97%

$40.86 - $586.00

Chemical Identifiers

CAS 30766-03-1
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD03426665
InChI Key RPHHYRNGCJYQSP-UHFFFAOYSA-N
Synonym 4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
PubChem CID 735150
IUPAC Name 4-bromopyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1Br)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6424403 View Documents Alfa Aesar™
H6424403
1g Each for $40.86
 
AAH6424406 View Documents Alfa Aesar™
H6424406
5g Each for $147.27
 
AAH6424414 View Documents Alfa Aesar™
H6424414
25g Each for $586.00
 
Description

Description

Applications
4-Bromopicolinic acid is a useful synthetic intermediate. Also used as a intermediate for agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in dimethyl sulfoxide , methanol and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

30766-03-1
202.007
RPHHYRNGCJYQSP-UHFFFAOYSA-N
735150
C1=CN=C(C=C1Br)C(=O)O
C6H4BrNO2
MFCD03426665
4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
4-bromopyridine-2-carboxylic acid

Specifications

177°C to 179°C
30766-03-1
MFCD03426665
4-bromopicolinic acid, 4-bromo-2-pyridinecarboxylic acid, 4-bromo-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 4-bromo, 4-bromo-pyridine-2-carboxylic acid, aurora 23242, pubchem4064, acmc-209hio, ksc222g5r
C1=CN=C(C=C1Br)C(=O)O
202.007
202.01
4-Bromopyridine-2-carboxylic acid
1g
C6H4BrNO2
471891
RPHHYRNGCJYQSP-UHFFFAOYSA-N
4-bromopyridine-2-carboxylic acid
735150
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning