missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromopyrene, TCI America™

$812.80 - $2,983.81

Chemical Identifiers

CAS 1732-26-9
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
InChI Key QMHTZTOPYZKQLC-UHFFFAOYSA-N
PubChem CID 3014036
IUPAC Name 4-bromopyrene
SMILES C1=CC2=C3C(=C1)C=C(C4=CC=CC(=C43)C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28071G View Documents TCI America
B28071G
1g Each for $812.80
 
B28075G View Documents TCI America
B28075G
5g Each for $2,983.81
 
Specifications

Chemical Identifiers

1732-26-9
281.152
3014036
C1=CC2=C3C(=C1)C=C(C4=CC=CC(=C43)C=C2)Br
C16H9Br
QMHTZTOPYZKQLC-UHFFFAOYSA-N
4-bromopyrene

Specifications

Yellow
1g
C16H9Br
C1=CC2=C3C(=C1)C=C(C4=CC=CC(=C43)C=C2)Br
281.152
281.15
≥95.0% (GC)
152°C
1732-26-9
QMHTZTOPYZKQLC-UHFFFAOYSA-N
4-bromopyrene
3014036
Crystal-Powder at 20°C
4-Bromopyrene
Safety and Handling

Safety and Handling

TSCA : No