missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromophthalimide, ≥97%

Manufacturer:  Alfa Aesar™ H6413603

 View more versions of this product

Catalog No. AAH6413603



Description

Description

Applications
It is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Soluble in N,N-DMF almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

4-Bromophthalimide
230°C to 235°C
6941-75-9
MFCD00466049
4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
C1=CC2=C(C=C1Br)C(=O)NC2=O
226.029
226.03
≥97%
White
1g
C8H4BrNO2
131235
GNYICZVGHULCHE-UHFFFAOYSA-N
5-bromoisoindole-1,3-dione
236018
Solid

Chemical Identifiers

6941-75-9
226.029
GNYICZVGHULCHE-UHFFFAOYSA-N
236018
C1=CC2=C(C=C1Br)C(=O)NC2=O
C8H4BrNO2
MFCD00466049
4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
5-bromoisoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents