missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophthalic Acid, TCI America™

$22.92 - $201.67

Chemical Identifiers

CAS 6968-28-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00043538
InChI Key AZXKGUVDIORSED-UHFFFAOYSA-N
Synonym 4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
PubChem CID 81428
IUPAC Name 4-bromophthalic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22571G View Documents TCI America
B22571G
1g Each for $22.92
 
B22575G View Documents TCI America
B22575G
5g Each for $61.11
 
B225725G View Documents TCI America
B225725G
25g Each for $201.67
 
Specifications

Chemical Identifiers

6968-28-1
245.028
AZXKGUVDIORSED-UHFFFAOYSA-N
81428
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
C8H5BrO4
MFCD00043538
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
4-bromophthalic acid

Specifications

Undesignated
1g
C8H5BrO4
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
245.028
245.03
≥98.0% (T)
166°C
6968-28-1
MFCD00043538
AZXKGUVDIORSED-UHFFFAOYSA-N
4-bromophthalic acid
81428
Crystal-Powder at 20°C
4-Bromophthalic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0062

TSCA : No