missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromophthalic acid, 98%

Manufacturer:  Alfa Aesar™ L1341803

 View more versions of this product

Catalog No. AAL1341803



Specifications

Specifications

4-Bromophthalic acid
1g
C8H5BrO4
1959086
AZXKGUVDIORSED-UHFFFAOYSA-N
4-bromophthalic acid
81428
98%
162°C to 164°C
6968-28-1
MFCD00043538
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
245.028
245.03

Chemical Identifiers

6968-28-1
245.028
AZXKGUVDIORSED-UHFFFAOYSA-N
81428
C1=CC(=C(C=C1Br)C(=O)O)C(=O)O
C8H5BrO4
MFCD00043538
4-bromophthalicacid, 4-bromobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-bromo, zlchem 539, 5-bromo-phthalic acid, acmc-209oah, #, 4-bromo-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 4-bromo, 4-bromobenzene-1,2-dioic acid
4-bromophthalic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents