missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5553414
Description
It is used as the intermediate of Organic Light Emitting Diode (OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as the intermediate of Organic Light Emitting Diode (OLED).
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
(4-Bromophenylethynyl)trimethylsilane | |
White to Yellow | |
Powder | |
C11H13BrSi | |
4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane | |
C[Si](C)(C)C#CC1=CC=C(Br)C=C1 | |
253.21 | |
253.21 |
61°C to 63°C | |
16116-78-2 | |
25 g | |
MFCD01863556 | |
RNMSGCJGNJYDNS-UHFFFAOYSA-N | |
2-(4-bromophenyl)ethynyl-trimethylsilane | |
4226980 | |
98% |
Chemical Identifiers
16116-78-2 | |
253.21 | |
RNMSGCJGNJYDNS-UHFFFAOYSA-N | |
4226980 | |
C[Si](C)(C)C#CC1=CC=C(Br)C=C1 |
C11H13BrSi | |
MFCD01863556 | |
4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane | |
2-(4-bromophenyl)ethynyl-trimethylsilane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only