Please login to your online account to display your discounted pricing

4-Bromophenylboronic acid, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC107280250

 View more versions of this product

Catalog No. AC107280250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 200.83
Quantity 25g
Color Off-white to light beige
Color Index No. 4-Bromophenylboronic acid
Beilstein 16, II, 638
CAS Max % 100.0
CAS 5467-74-3
Linear Formula BrC6H4B(OH)2
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 5467-74-3
Health Hazard 2 GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Molecular Formula C6H6BBrO2
CAS Min % 97.5
MDL Number MFCD00002104
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Chemical Name or Material 4-Bromophenylboronic acid

Specifications

Chemical Name or Material 4-Bromophenylboronic acid
CAS 5467-74-3
CAS Min % 97.5
CAS Max % 100.0
CAS 5467-74-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6BBrO2
Linear Formula BrC6H4B(OH)2
Formula Weight 200.83
MDL Number MFCD00002104
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.

SDS & Safety Documents