Please login to your online account to display your discounted pricing

4-Bromophenylacetone, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC380220010

 View more versions of this product

Catalog No. AC380220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 213.08
Quantity 1g
Color Clear yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Bromophenylacetone
Refractive Index 1.5560 to 1.5590 (20°C, 589nm)
Molecular Formula C9H9BrO
MDL Number MFCD00210401
CAS 6186-22-7
Assay Percent Range 97%
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Bromophenylacetone
CAS 6186-22-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9BrO
Formula Weight 213.08
MDL Number MFCD00210401