missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenylacetic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 160520050

 View more versions of this product

Catalog No. AC160520050



Specifications

Specifications

4-Bromophenylacetic acid
Glass bottle
112°C to 114°C
1878-68-8
100.0
C8H7BrO2
MFCD00004342
2-4-bromophenyl acetic acid
QOWSWEBLNVACCL-UHFFFAOYSA-N
2-(4-bromophenyl)acetic acid
74654
215.05
99%
Authentic
Undesignated
5g
98.5
99%
BrC6H4CH2CO2H
09, 451
Solubility in water: slightly soluble in water. Other solubilities: very soluble in ethyl alcohol,diethyl ether,carb
C1=CC(=CC=C1CC(=O)O)Br
215.05
CHEBI:1790
Crystalline Powder

Chemical Identifiers

1878-68-8
215.05
QOWSWEBLNVACCL-UHFFFAOYSA-N
74654
2-(4-bromophenyl)acetic acid
C8H7BrO2
MFCD00004342
2-4-bromophenyl acetic acid
CHEBI:1790
C1=CC(=CC=C1CC(=O)O)Br
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-523-7

SDS Documents