missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1110114

 View more versions of this product

Catalog No. AAL1110114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromophenyl isothiocyanate
144°C to 145°C (5mmHg)
25g
C7H4BrNS
UN1759
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
C1=CC(=CC=C1N=C=S)Br
214.08
214.09
>110°C (230°F)
56°C to 60°C
1985-12-2
MFCD00004808
878549
XQACWEBGSZBLRG-UHFFFAOYSA-N
1-bromo-4-isothiocyanatobenzene
16133
97%

Chemical Identifiers

1985-12-2
214.08
XQACWEBGSZBLRG-UHFFFAOYSA-N
16133
C1=CC(=CC=C1N=C=S)Br
C7H4BrNS
MFCD00004808
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
1-bromo-4-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

RTECSNumber : NX8400000

Documents