missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Bromophenyl)cyclopropylmethanone, 95%, ACROS Organics™

Chemical Identifiers

CAS 6952-89-2
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD00019227
InChI Key QTHHOINSCNBYQO-UHFFFAOYSA-N
Synonym 4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromophenyl cyclopropyl ketone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
PubChem CID 81394
IUPAC Name (4-bromophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC235422500 SDS View Documents Acros Organics
AC235422500
250g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC235420500 SDS View Documents Acros Organics
AC235420500
50g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6952-89-2
225.085
QTHHOINSCNBYQO-UHFFFAOYSA-N
81394
C1CC1C(=O)C2=CC=C(C=C2)Br
C10H9BrO
MFCD00019227
4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromophenyl cyclopropyl ketone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
(4-bromophenyl)-cyclopropylmethanone

Specifications

1.4500g/mL
Glass bottle
250g
94.0
95%
MFCD00019227
QTHHOINSCNBYQO-UHFFFAOYSA-N
(4-bromophenyl)-cyclopropylmethanone
81394
Crystalline Powder or Low Melting Crystalline Mass
(4-Bromophenyl)cyclopropylmethanone
Authentic
Beige to Orange or Yellow
6952-89-2
100.0
C10H9BrO
4-bromophenyl cyclopropyl methanone, 4-bromophenyl cyclopropyl ketone, 4-bromophenyl cyclopropylmethanone, methanone, 4-bromophenyl cyclopropyl, 4-bromophenyl cyclopropyl ketone, methanone, 4-bromophenyl cyclopropyl, 4-bromo-cyclopropylcarbonylbenzene, acmc-1bag8, 4-bromobenzoyl cyclopropane, bromophenylcyclopropylmethanone
C1CC1C(=O)C2=CC=C(C=C2)Br
225.085
225.09
≥94%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.