missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenyl chloroformate, +95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC430150050

 View more versions of this product

Catalog No. AC430150050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥95.0%
235.46
1.6480g/mL
100.0
C7H4BrClO2
4-Bromophenyl chloroformate
Glass bottle
Low Melting Solid or Liquid
5g
95.0
MFCD00013256
7693-44-9
Authentic
95+%

Chemical Identifiers

7693-44-9
100.0
C7H4BrClO2
MFCD00013256
≥95.0%
95.0
95+%
235.46
4-Bromophenyl chloroformate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger