missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenyl acetate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452770050

 View more versions of this product

Catalog No. AC452770050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromophenyl acetate
1927-95-3
C8H7BrO2
XEXHCQJRVMVJMY-UHFFFAOYSA-N
(4-bromophenyl) acetate
74726
99%
5g
98.5% min. (GC)
4-bromoacetoxybenzene, 4-bromophenol acetate, 4-bromophenyl acetate, acetic acid 4-bromophenyl ester, p-acetoxybromobenzene, p-bromophenyl acetate, phenol, 4-bromo-, 1-acetate, phenol, 4-bromo-, acetate, phenol, 4-bromo-,1-acetate, phenol, p-bromo-, acetate
CC(=O)OC1=CC=C(C=C1)Br
215.05
215.05

Chemical Identifiers

1927-95-3
215.05
4-bromoacetoxybenzene, 4-bromophenol acetate, 4-bromophenyl acetate, acetic acid 4-bromophenyl ester, p-acetoxybromobenzene, p-bromophenyl acetate, phenol, 4-bromo-, 1-acetate, phenol, 4-bromo-, acetate, phenol, 4-bromo-,1-acetate, phenol, p-bromo-, acetate
(4-bromophenyl) acetate
C8H7BrO2
XEXHCQJRVMVJMY-UHFFFAOYSA-N
74726
CC(=O)OC1=CC=C(C=C1)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
Avoid release

GHS Signal Word: Warning

EINECSNumber : 217-668-6

SDS Documents