missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromophenyl acetate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452770250
Specifications
4-Bromophenyl acetate | |
1927-95-3 | |
C8H7BrO2 | |
XEXHCQJRVMVJMY-UHFFFAOYSA-N | |
(4-bromophenyl) acetate | |
74726 | |
99% |
25g | |
98.5% min. (GC) | |
4-bromoacetoxybenzene, 4-bromophenol acetate, 4-bromophenyl acetate, acetic acid 4-bromophenyl ester, p-acetoxybromobenzene, p-bromophenyl acetate, phenol, 4-bromo-, 1-acetate, phenol, 4-bromo-, acetate, phenol, 4-bromo-,1-acetate, phenol, p-bromo-, acetate | |
CC(=O)OC1=CC=C(C=C1)Br | |
215.05 | |
215.05 |
Chemical Identifiers
1927-95-3 | |
215.05 | |
4-bromoacetoxybenzene, 4-bromophenol acetate, 4-bromophenyl acetate, acetic acid 4-bromophenyl ester, p-acetoxybromobenzene, p-bromophenyl acetate, phenol, 4-bromo-, 1-acetate, phenol, 4-bromo-, acetate, phenol, 4-bromo-,1-acetate, phenol, p-bromo-, acetate | |
(4-bromophenyl) acetate |
C8H7BrO2 | |
XEXHCQJRVMVJMY-UHFFFAOYSA-N | |
74726 | |
CC(=O)OC1=CC=C(C=C1)Br |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
Avoid release
GHS Signal Word: Warning
EINECSNumber : 217-668-6