missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenoxyacetic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6088814

 View more versions of this product

Catalog No. AAH6088814



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Bromophenoxyacetic acid
149°C to 153°C
MFCD00093070
4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy
C1=CC(=CC=C1OCC(=O)O)Br
231.045
231.04
1878-91-7
C8H7BrO3
25 g
SZEBGAQWWSUOHT-UHFFFAOYSA-N
2-(4-bromophenoxy)acetic acid
74657
95%

Chemical Identifiers

1878-91-7
231.045
SZEBGAQWWSUOHT-UHFFFAOYSA-N
74657
C1=CC(=CC=C1OCC(=O)O)Br
C8H7BrO3
MFCD00093070
4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy
2-(4-bromophenoxy)acetic acid
Safety and Handling

Safety and Handling

EINECSNumber : 217-530-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions