Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Bromophenoxy)trimethylsilane, 98%

$53.04 - $640.38

Chemical Identifiers

Chemical Name or Material (4-Bromophenoxy)trimethylsilane
CAS 17878-44-3
Molecular Formula C9H13BrOSi
Formula Weight 245.2
MDL Number MFCD00040713
Synonym 1-Bromo-4-(trimethylsiloxy)benzene; 4-(Trimethylsiloxy)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2495606

  SDS
alfa aesar™
B2495606
5g Each for $53.04

AAB2495614

  SDS
alfa aesar™
B2495614
25g Each for $203.42

AAB2495622

  SDS
alfa aesar™
B2495622
100g Each for $640.38
Description & Specifications

Specifications

Formula Weight 245.2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
TSCA TSCA
Molecular Formula C9H13BrOSi
MDL Number MFCD00040713
Flash Point 97°C (207°F)
Chemical Name or Material (4-Bromophenoxy)trimethylsilane
Health Hazard 1 Warning
Synonym 1-Bromo-4-(trimethylsiloxy)benzene; 4-(Trimethylsiloxy)bromobenzene
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 17878-44-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.