Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromophenetole, 98%

$33.30 - $113.01

Chemical Identifiers

Chemical Name or Material 4-Bromophenetole
CAS 588-96-5
Molecular Formula C8H9BrO
Formula Weight 201.06
MDL Number MFCD00000098
Beilstein 2042066
Merck Index 14,1428
Synonym 4-Bromoethoxybenzene; 4-Bromophenyl ethyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2139814

  SDS
alfa aesar™
B2139814
25g Each for $33.30

AAB2139822

  SDS
alfa aesar™
B2139822
100g Each for $113.01
Description & Specifications

Specifications

Formula Weight 201.06
Chemical Name or Material 4-Bromophenetole
TSCA TSCA
Boiling Point 232° to 234°C
Melting Point 3° to 5°C
Quantity 25g
Flash Point 103°C (217°F)
Assay 98%
Molecular Formula C8H9BrO
MDL Number MFCD00000098
CAS 588-96-5
Beilstein 2042066
Merck Index 14,1428
Synonym 4-Bromoethoxybenzene; 4-Bromophenyl ethyl ether